Learn More
Eppendorf™ Sandwich-Mode Spacer
Required for stacking rotors F-45-72-8 and F-45-48-11
Brand:  Eppendorf™ 5301316005
Product Code. 10397083
  • £29.10 / Each
Explore more special offers
Description
Description
  • Rotor spacer
  • For use with rotors F-45-72-8 and F-45-48-11
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Spacer
Rotors F-45-72-8 and F-45-48-11
Required for stacking rotors F-45-72-8 and F-45-48-11