Learn More
Applied Biosystems™ Sanger Sequencing Kit
Sanger Sequencing Kit includes all reagents needed for sample preparation for sanger sequencing
Brand: Applied Biosystems™ A38073
743.00 GBP valid until 2025-03-28
Use promo code "24107" to get your promotional price.
Includes: 2 x 100 rxns ExoSAP-IT Express, 2 x BigDye Terminator v1.1 and v3.1 sequencing buffer (5X), 1 x 800 μL BigDye Terminator v3.1 ready reaction mix, 1 tube M13 (-21) primer, 1 tube pGEM control DNA, 1 x 2 mL XTerminator solution, 1 x 9 mL SAM solution
Description

Specifications
• 2 x 100 rxns ExoSAP-IT Express, store at -5 to -30°C • 2 x BigDye Terminator v1.1 and v3.1 sequencing buffer (5X), store at 2–8°C • 1 x 800 µL BigDye Terminator v3.1 ready reaction mix, store at -15 to -25°C • 1 tube M13 (-21) primer, store at -15 to -25°C • 1 tube pGEM control DNA, store at -15 to -25°C • 1 x 2 mL XTerminator solution, store at 2–8°C • 1 x 9 mL SAM solution, store at room temperature or 4°C |
|
Sequencing Instruments (Capillary Electrophoresis-Based) | |
200 | |
200 Reactions |
Kit | |
Standard Sequencing | |
Sanger Sequencing Kit | |
Approved for shipment at Room Temperature or on Wet or Dry Ice |
Safety and Handling
- Sanger Sequencing Kit
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : benzene
Special Offers
- Multiple Manufacturers
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.