Learn More
Notrax Sani-Trax™ Disinfecting Foot Bath Entrance Mat, Black
Notrax Sani-Trax™ Disinfecting Foot Bath Entrance Mat cleans and sanitizes shoes, recommended at entrances to all food processing areas. As traffic passes, rubber tips bend to immerse soles in disinfectant solution, a double action that keeps contamination from reaching food processing areas.
£39.40 - £55.55
Specifications
Color Black
Material Natural Rubber
Type Industrial and Commercial Matting
View More Specs
Products 2
Product Code Brand Length (Metric) Width (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Width (Metric) Price Quantity & Availability  
17782018
View Documents
Notrax
346S1824BL
24 cm 18 cm
£39.40
Each
Estimated Shipment: 11-04-2025
to see stock.
Add to basket
17792018
View Documents
Notrax
346S2432BL
80 cm 60 cm
£55.55
Each
Estimated Shipment: 11-04-2025
to see stock.
Add to basket
Description
Description
  • This mat cleans and sanitizes shoes, recommended at entrances to all food processing areas
  • Thousands of flexible fingers provide an automatic scrubbing action that dislodges contaminants from footwear
  • As traffic passes, rubber tips bend under pressure to immerse soles in disinfectant solution, a double action that keeps contamination from reaching food processing areas
  • Convenient mat size fits most doorways
  • Suction cups on underside prevents mat from shifting
Specifications
Specifications
Black
Natural Rubber
Industrial and Commercial Matting
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Notrax Sani-Trax™ Disinfecting Foot Bath Entrance Mat, Black