Learn More
Masterflex™ Sanitary Clamp to Hose Barb Straight Adapter Fittings
Maintain sanitary conditions
£106.00 - £134.00
Specifications
Product Type | Sanitary Barbed Adapter |
---|---|
Material | Polypropylene |
Packaging Quantity | 6 |
Hose Barb | 1/4 in. |
Product Code | Brand | Hose Barb | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Hose Barb | Includes | Price | Quantity & Availability | ||||
15956982
|
Masterflex™
31200-39 |
1/4 in. | Mini Tri-Clamp |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
16458205
|
Masterflex™
MK-31200-44 |
1/4 in. | 1.5 in. Tri-Clamp |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Meet requirements for USP Class VI, FDA, REACH, and California Proposition 65
- Surface finishes to 15 micron minimizes adhesion or entrapment
- Packaged to exacting tolerances under ISO Class 7 cleanroom conditions
- Compatible with other Tri-Clamp™ connections
- The sanitary Tri-Clamp barbed adapters are ideal for use in the biopharmaceutical, food and beverage, and single-use disposable industries.
- The fittings are made from animal-derived ingredient-free and phthalate-free polypropylene (PP)
- They have excellent chemical resistance and may be sterilized by gamma irradiation, EtO, E-beam, or autoclave
Specifications
Sanitary Barbed Adapter | |
Polypropylene | |
6 | |
1/4 in. |