Learn More
Cole-Parmer™ Sanitary Coupling with Barb, PVDF
Cole-Parmer™ Sanitary Coupling with Barb, PVDF has a tapered funnel design to provide a steady fluid flow.
Brand: Cole-Parmer™ 31512-04
This item is not returnable.
View return policy
Description
- Reduces dead space and improves flow.
- Tapered funnel design provides a steady fluid flow.
- Smooth inner surface prevents particle entrapment.
- Seamless construction prevents contamination by eliminating leaks and areas of potential bacterial growth.
- Material meets USP Class VI requirements and is FDA compliant for food contact.
Specifications
Meets USP Class VI requirements and is FDA compliant for food contact | |
PVDF | |
FDA compliant for food contact | |
Gamma | |
1 11/16 in. |
Not Autoclavable | |
3/8 in. ID tubing, 1/2 in. mini clamp | |
Fits 3/8 in. ID tubing | |
31/32 in. |