missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Sapphire PCR Tubes with Attached Frosted Flat Cap

Product Code. 13025063
Click to view available options
Color:
Assorted
Blue
Green
Natural
Red
Violet
Yellow
Volume (Metric):
0.2 mL
0.5 mL
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 13025063

Brand: Greiner Bio-One 683275

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For use in RT/Quantitative-PCR. Greiner Bio-One Sapphire PCR Tubes with Attached Frosted Flat Cap are comprehensive thermal cycler compatible.

  • Thin-walled for heat transfer during amplification
  • Free of detectable DNase, RNase, human DNA, endotoxin and PCR inhibitors
  • Packaging labelled with product symbols for warehousing
  • Used for sample handling
TRUSTED_SUSTAINABILITY

Specifications

Color Green
Volume (Metric) 0.2 mL
For Use With (Equipment) PCR
Sterility Non-sterile
Material Polypropylene
Closure Type Attached Frosted Flat Cap
Product Type PCR Tubes with Attached Flat Caps
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.