missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Sapphire RT-PCR Breakable 8-Tube Strips with Individual Attached Caps

For use with real-time/quantitative PCR. Greiner Bio-One Sapphire RT-PCR Breakable 8-Tube Strips with Individual Attached Caps are compatible with comprehensive thermal cycler.

£168.30

Specifications

Color Clear
Volume (Metric) 0.2 mL
For Use With (Equipment) Real-time/quantitative PCR
Sterility Non-sterile
Material Polypropylene
View More Specs

Products 1
Product Code Brand Closure Type Price Quantity & Availability  
Product Code Brand Closure Type Price Quantity & Availability  
17149268
View Documents Promotion Details
Greiner Bio-One
673284
Optical (Individually Attached) This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

  • Thin-walled for heat transfer during amplification
  • Free of detectable DNase, RNase, human DNA, endotoxin and PCR inhibitors
  • Packaging labelled with product symbols for warehousing
  • Used for sample handling
Specifications

Specifications

Clear
Real-time/quantitative PCR
Polypropylene
0.2 mL
Non-sterile
RT-PCR 8-Tube Strips
Videos
SDS
Documents

Documents

Product Certifications