missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sapphire substrate, 50.8mm dia x 0.432mm thick, EPI polished one side, C-axis, HEMCOR single crystal

£123.00 - £582.00

Specifications

Product Type Sapphire Substrate, HEMCOR Single Crystal
Description EPI polished one side
Diameter (Metric) 50.8 mm
Thickness 0.432 mm
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15443925
View Documents
Thermo Scientific Chemicals
045019.KT
1 Ea.
£123.00
Each
In Stock
to see stock.
Add to basket
 
15453925
View Documents
Thermo Scientific Chemicals
045019.KU
5 Ea.
£582.00
Pack of 5
In Stock
to see stock.
Add to basket
 
Description

Description

Sapphire is used as an insulating substrate upon which to deposit silicon to make the integrated circuits, high-power radio-frequency (RF) applications such as those found in cellular telephones, public-safety band radios, and satellite communication systems, used in the semiconductor industry, blue light-emitting diodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sapphire Substrate, HEMCOR Single Crystal
EPI polished one side
50.8 mm
0.432 mm
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.