missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAPS1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | SAPS1 |
---|---|
Concentration | 0.29 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16837814
|
Proteintech
17819-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16827814
|
Proteintech
17819-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Protein phosphatase regulatory subunits, such as SAPS1, modulate the activity of protein phosphatase catalytic subunits by restricting substrate specificity, recruiting substrates, and determining the intracellular localization of the holoenzyme. SAPS1 is a regulatory subunit for the protein phosphatase-6 catalytic subunit.Specifications
SAPS1 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
Q7TSI3, Q9UPN7 | |
22870, 243819 | |
SAPS1 Fusion Protein Ag12149 | |
Primary | |
-20°C | |
PPP6R1 |
0.29 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
KIAA1115, PP6R1, PPP6R1, SAP190, SAPS domain family member 1, SAPS domain family, member 1, SAPS1 | |
PPP6R1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |