missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sarcosine methyl ester hydrochloride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
It can react with formic acid to get N-formyl-N-methyl-glycine methyl ester in the presence of HCO2Na at ambient temperature.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It can react with formic acid to get N-formyl-N-methyl-glycine methyl ester in the presence of HCO2Na at ambient temperature.
Solubility
Soluble in water.
Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
It can react with formic acid to get N-formyl-N-methyl-glycine methyl ester in the presence of HCO2Na at ambient temperature.
Solubility
Soluble in water.
Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | ∽105°C (decomposition) |
Quantity | 10 g |
Beilstein | 3677128 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 139.58 |
Percent Purity | 98% |
Chemical Name or Material | Sarcosine methyl ester hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'