missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ SARS-CoV-2 Envelope Protein (YP_009724392.1) VersaClone cDNA
Contains gene inserts that are fully sequenced and produced with high quality
Brand: R&D Systems™ RDC3127
Additional Details : Weight : 0.00970kg
Specifications
Envelope | |
10 μg at 0.2 μg/μL | |
241 bp | |
SARS-CoV-2 |
YP_009724392.1 | |
cDNA is provided in 10 mM Tris-Cl, pH 8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |