Learn More
SARS-CoV-2 nsp13 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Bio-Techne NBP3-07055FR
Product Code. 18318093
  • £516.00 / 0.10mL
Explore more special offers
Description
Description
SARS-CoV-2 nsp13 Polyclonal antibody specifically detects SARS-CoV-2 nsp13 in SARS-CoV-2 samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence
Specifications
Specifications
SARS-CoV-2 nsp13
Polyclonal
50mM Sodium Borate with 0.05% Sodium Azide
Rabbit
Peptide Affinity-purified
RUO
Primary
SARS-CoV-2
Purified
ELISA, Immunohistochemistry, Immunofluorescence
DyLight 680
helicase, Non-structural protein 13
This antibody was raised against a peptide corresponding to 19 amino acids near the carboxyl terminus of the SARS-CoV-2 (COVID-19) NSP13 (Helicase) protein. The immunogen is located in the last 50 amino acids of SARS-CoV-2 (COVID-19) NSP13 (Helicase).
0.1 mL
Infections (Virus Bacteria and Parasites)
43740578
Store at 4°C in the dark.