Learn More
IBA Lifesciences SARS-CoV-2 Peptide KIADYNYKL (HLA-A*0201)
GREENER_CHOICE
Single peptide synthesized as presented by the MHC class I allele
Brand:  IBA Lifesciences 6-7119-901
Product Code. 16604754
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • SARS-CoV-2 peptide KIADYNYKL (HLA-A*0201) is a single peptide for stimulation of T cells.
  • The peptide from SARS-CoV-2 is synthesized as it is presented by the MHC class I HLA-A*0201 allele and can be used for targeted in vitro generation and expansion of antigen-specific CD8+ T cells.
  • The peptides were purified by HPLC using a water and acetonitrile gradient and are endotoxin-free.
Specifications
Specifications
>85%
Lyophilized Powder
T cell stimulation
Free