Learn More
IBA Lifesciences SARS-CoV-2 Peptide LALLLLDRL (HLA-A*0201)
Single peptide synthesized as presented by the MHC class I allele
Brand: IBA Lifesciences 6-7116-901
This item is not returnable.
View return policy
Description
- SARS-CoV-2 peptide ILLNKHIDA (HLA-A*0201) is a single peptide for stimulation of T cells.
- The peptide from SARS-CoV-2 is synthesized as it is presented by the MHC class I HLA-A*0201 allele and can be used for targeted in vitro generation and expansion of antigen-specific CD8+ T cells.
- The peptides were purified by HPLC using a water and acetonitrile gradient and are endotoxin-free.
Specifications
>85% | |
Lyophilized Powder | |
T cell stimulation |
Free | |
1 mg |