Learn More
SARS-CoV-2 Spike Human anti-Virus, Unconjugated, Clone: AM038105, Proteintech
Human Recombinant Antibody
Brand: Proteintech 91339-PTG-100UG
This item is not returnable.
View return policy
Description
The SARS-CoV-2 Spike antibody from Proteintech is a human recombinant antibody generated with virus of virus SARS-CoV-2 Spike. This antibody recognizes virus antigen. The SARS-CoV-2 Spike antibody has been validated for the following applications: ELISA analysis.
Specifications
SARS-CoV-2 Spike | |
Monoclonal | |
Unconjugated | |
100 μg | |
43740568 | |
IgG1 |
ELISA | |
AM038105 | |
Human | |
Primary | |
Virus |