missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 Spike Human anti-Virus, Unconjugated, Clone: AM038105, Proteintech
Human Recombinant Antibody
£177.00 - £530.00
Specifications
Antigen | SARS-CoV-2 Spike |
---|---|
Clone | AM038105 |
Applications | ELISA |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16890436
|
Proteintech
91339-PTG-20UG |
20 μg |
£177.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16880436
|
Proteintech
91339-PTG-100UG |
100 μg |
£530.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SARS-CoV-2 Spike antibody from Proteintech is a human recombinant antibody generated with virus of virus SARS-CoV-2 Spike. This antibody recognizes virus antigen. The SARS-CoV-2 Spike antibody has been validated for the following applications: ELISA analysis.
Specifications
SARS-CoV-2 Spike | |
ELISA | |
Unconjugated | |
Virus | |
IgG1 |
AM038105 | |
Monoclonal | |
Human | |
43740568 | |
Primary |