missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV/SARS-CoV-2 NSP8 Monoclonal Antibody (5A10), Invitrogen™
Mouse Monoclonal Antibody
Brand: Thermo Scientific MA535947
416.02 GBP valid until 2025-03-29
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
The nonstructural protein 8 (nsp8) is one of the SARS-Coronavirus replicase cleaving products, encoded by ORF1a. Nsp8 is thought to be part of the viral replication complex, which is associated with intracellular membranes. No specific information on the function of nsp8 is available.
Specifications
SARS-CoV/SARS-CoV-2 NSP8 | |
Monoclonal | |
0.67 mg/mL | |
PBS with no preservative | |
P0DTD1 | |
orf1ab | |
SARS-CoV NSP8 | |
100 μL | |
Primary | |
Virus | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Western Blot | |
5A10 | |
Unconjugated | |
orf1ab | |
COVID-19; COVID-19 Envelope; SARS-CoV-2; SARS-CoV-2 Envelope | |
Mouse | |
Protein G | |
RUO | |
43740578 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SARS-CoV/SARS-CoV-2 NSP8 Monoclonal Antibody (5A10), Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction