missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS Nucleocapsid Protein Antibody Pack, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05695
Additional Details : Weight : 0.58200kg
Description
SARS Nucleocapsid Protein Polyclonal antibody specifically detects SARS Nucleocapsid Protein in SARS-CoV, SARS-CoV-2 samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
SARS Nucleocapsid Protein | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
2019-nCoV Nucleocapsid Protein, COVID-19 N protein, COVID-19 nucleocapsid protein, N, N protein, N structural protein, NC, Nucleocapsid protein, Nucleoprotein, Protein N, SARS coronavirus N protein, SARS coronavirus nucleocapsid protein, SARS CoV, SARS CoV N protein, SARS CoV nucleocapsid protein, SARS N protein, SARS Nucleoprotein, SARSCoV, SARSCoV N protein, SARSCoV nucleocapsid protein, SARS-CoV-2 N protein, SARSCOV2 N protein, SARS-COV-2 nucleocapsid protein, SARS-COV-2 Nucleoprotein, Severe acute respiratory syndrome | |
See individual datasheets. | |
RUO | |
1489678 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
NB100-56576 and NB100-56683: PBS, 0.05% Sodium Azide. NBP2-44205: 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% Sodium Azide. | |
Rabbit | |
3 Vials | |
Primary | |
SARS-CoV, SARS-CoV-2 |