missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Sartobind Lab Affinity IDA75

Ready-to-use membrane adsorbers for immobilized metal affinity chromatography (IMAC) in syringe filter format. Superior flow rates and high binding capacity based on the patented macroporous Sartobind™ membranes with iminodiacetic acid (IDA).

Brand:  Sartorius 93IDA-42DB-12--V

Product Code. 11740926

  • £425.00 / Pack of 2

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Based on the Patented Sartobind™ Platform Sartobind™ Lab membrane adsorbers for immobilized metal affinity chromatography (IMAC) are ready-to-use units in a syringe filter format.
  • They are based on the patented macroporous Sartobind™ membranes, with iminodiacetic acid (IDA) covalently bound to the entire internal surface of the pores. This results in a chromatography matrix with superior flow rates and high binding capacity.
  • The IDA ligand enables coupling of the metal ion of your choice to the membrane adsorber, ensuring simple process optimization for maximum purity and yield of each his-tagged protein.
  • Ultimate Versatility Sartobind™ Lab IDA units are available with a 2.1 mL bed volume, providing binding capacities up to 7.5 mg his-tagged protein per unit.
  • Equipped with standard Luer lock connectors, Sartobind™ Lab units enable simple, equipment-free handling by syringe, or can be easily connected to FPLC systems for automated purifications.
  • The units are compatible with most common chromatography buffers and can be regenerated for multiple purification cycles, ensuring high-cost efficiency.
  • Fewer preparative steps
  • Greatly reduced cycle times
  • Achieve higher yields in less time
  • Purify equipment-free or with an FPLC system
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2.1 mL
Reusable
Syringe
Cellulose
Non-sterile
Polysulfone
0.6 mPa
Luer Male
1.3 mL
Air Syringe Filter
Polyester
5.0 μm
His-tagged Protein Purification
Luer Female
Metal Chelate
2Pack
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers