missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Sartobind Lab Affinity Pr A

Sartobind™ Lab units are available with a 2 mL bed volume, providing binding capacities of 10 to 15 mg IgG per unit. Sartobind™ Lab units enable simple equipment-free handling by syringe or can be easily connected to FPLC systems for automated purifications.

Brand:  Sartorius 93PRAP06HB-12--A


Product Code. 11770895

  • £899.00 / Pack of 4

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • High-Speed Purification Sartobind™ Lab Protein A affinity membrane adsorbers are ready-to-use units in a syringe filter format.
  • The macorporous matrix makes them the ideal tool for rapid purification of immunoglobulins (IgG) and monoclonal antibodies (mAbs) by providing superior flow rates and reduced cycle times when compared to conventional columns.
  • A complete purification cycle, whether by syringe or on an FPLC system, takes less than 5 minutes.
  • Ultimate Versatility Sartobind™ Lab Protein A units are available with a 2 mL bed volume, providing binding capacities of 10 to 15 mg IgG per unit. Equipped with standard Luer lock connectors, Sartobind™ Lab units enable simple, equipment-free handling by syringe, or can be easily connected to FPLC systems for automated purifications.
  • The units are compatible with most common protein A purification buffers and can be regenerated for multiple purification cycles, ensuring high-cost efficiency.
  • Greatly reduced cycle times
  • Achieve higher yields in less time
  • Purify equipment-free or with an FPLC system
  • Specifications:
    • Ligand: Protein A
    • Reference Protein: Human polyclonal IgG
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2.1 mL
Air Syringe Filter
Polyester
0.45 μm
mAb Purification
Luer Female
Luer Male
Membrane Adsorber
Reusable
Syringe
Cellulose
Non-sterile
Polypropylene
Affinity
4Pack
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • SARTOBIND PROTEIN A

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H228-Flammable solid.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Nitrocellulose

SDS
Documents

Documents

Certificates
Special Offers

Special Offers