missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Sartobind Lab IEX Q15
BIOPHARMA

Sartobind™ Lab units are available with strong or weak anion (Q and D) and strong cation (S) exchangers in three bed volumes ranging from 0.41 to 2.8 mL. Sartobind™ Lab units enable simple equipment-free handling by syringe or can be easily connected to FPLC systems for automated purifications.

Brand:  Sartorius 93IEXQ42GB-12--A

Product Code. 15235374

  • £235.00 / Pack of 4
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Process-Ready Sartobind™ Membrane Adsorbers Sartobind™ Lab membrane adsorbers for ion exchange chromatography are ready-to-use units in a syringe filter format.
  • They are based on the patented macroporous Sartobind™ membranes, with anion or cation exchange ligands covalently bound to the entire internal surface of the pores. This results in a media chromatography matrix with superior flow rates and high binding capacity.
  • Covalent linkage of ion exchange ligands to the membrane is very stable, allowing for many cycles of re-use without loss of binding capacity. In addition, these units utilize the same membrane adsorber platform as found in Sartobind™ capsules and cassettes, making them ideal for research and development work, prior to convenient scale-up for manufacturing.
  • Ultimate Versatility Sartobind™ Lab units for ion exchange are available with strong or weak anion (Q and D) and strong cation (S) exchangers, and in three bed volumes, ranging from 0.41 to 2.8 mL, corresponding to dynamic binding capacities between 12 and 80 mg protein per unit.
  • Equipped with standard Luer lock connectors, Sartobind™ Lab units enable simple, equipment-free handling by syringe, or can be easily connected to FPLC systems for automated purifications. The units are compatible with most common chromatography buffers and can be re-used 1,000 times, ensuring high-cost efficiency.
  • Greatly reduced cycle times
  • Achieve higher yields in less time
  • Purify equipment-free or with an FPLC system
  • Specifications: 
    • Ligand: Quaternary Ammonium
    • Reference Protein: BSA

TRUSTED_SUSTAINABILITY
Specifications

Specifications

2.8 mL
5.7 mL
Air Syringe Filter
Polyester
5 μm
Purification, Screening
Membrane Adsorber
Luer Lock
Reusable
Syringe
Cellulose
Non-sterile
Strong Anion Exchanger
66 x 31 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers