Learn More
Sartorius Sartolab™ MultiStation
Sartolab™ MultiStation is a stand specially designed to hold 1 to 6 vacuum filtration units, allowing simultaneous filtration of up to 6 samples.
Brand: Sartorius SDLC01
2606.71 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.
This item is not returnable.
View return policy
Description
- For hands-free parallel filtration of up to 6 samples
- Permanently connected to vacuum source
- Easily install vacuum filtration units in the MultiStation for quick and easy filtration of samples without the need for installation of extra connectors and time-consuming stabilization
- Works with Sartolab™ RF 50 vacuum filtration units; the funnel adapter of these units is designed to fit perfectly in the bracket of the MultiStation
- With one click, connect the filtration unit to the device, assuring perfect filter stability
- With a second click, engage the vacuum automatically and begin filtering
- Easily manipulate samples with the rotating, multi-directional head
- Easily keep track of samples during filtration with the numbered brackets
- Time-saving (no installation time for each filter unit before use)
Specifications
Not Autoclavable | |
Reusable | |
Sartolab RF/BT Units | |
Parallel filtration of up to 6 samples |
White | |
Vacuum Filter | |
Non-sterile | |
Filtration Unit |