missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Sartolab™ Single-Use Polystyrene Storage Bottle
Sartorius Sartolab single-use polystyrene bottles for storage of media, buffers, and other aqueous solutions.
Brand: Sartorius 180-22---------E
57.23 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- The bottles can be used with all Sartolab™ RF | BT vacuum filtration units with a 45 mm neck size
Specifications
Single-use | |
Polystyrene | |
150 mL |
Sartolab | |
Polystyrene | |
Sterile |