missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Sartolab™ Single-Use Polystyrene Storage Bottle

Sartorius Sartolab single-use polystyrene bottles for storage of media, buffers, and other aqueous solutions.

Brand:  Sartorius 180-24---------E

85.73 GBP valid until 2024-12-31
Use promo code "23270" to get your promotional price.



Additional Details : Weight : 1.71000kg

 View more versions of this product

Product Code. 17424412

  • £99.95 / Pack of 12
Estimated Shipment: 19-09-2024
to see stock.
Add to basket

Description

Description

  • The bottles can be used with all Sartolab™ RF | BT vacuum filtration units with a 45 mm neck size
Specifications

Specifications

Single-use
Polystyrene
500 mL
Sartolab
Polystyrene
Sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers