missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Sartolab™ Single-Use Polystyrene Storage Bottle
GREENER_CHOICEBIOPHARMA

Product Code. 17434412 Shop All Sartorius Products
Click to view available options
Sample Volume:
150 mL
250 mL
500 mL
1000 mL
Unit Size:
Pack of 12
This item is not returnable. View return policy

Product Code. 17434412

Brand: Sartorius undefined

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Sartorius Sartolab single-use polystyrene bottles for storage of media, buffers, and other aqueous solutions.

  • The bottles can be used with all Sartolab™ RF | BT vacuum filtration units with a 45 mm neck size
TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
For Use With (Equipment) Sartolab
Material Polystyrene
Material (Membrane) Polystyrene
Sample Volume 1000 mL
Sterility Sterile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.