Learn More
Sartorius arium™ Foot Switch
More Convenient Pure Water Dispensing
Brand: Sartorius H2O-AFS1
Code : NEW
Additional Details : Weight : 10.00000kg
Description
For external control of the time / volume dispense port for arium 611 systems.
- Water dispensing at a press of the foot
- Ideally suited for clean rooms, minimize the risk of contamination
- Comfortable fatigue-free switching thanks to low height
Specifications
Arium Water Purification System | |
Foot Switch |