Learn More
Sartorius arium™ Pro ASTM TYPE 1 - Ultrapure Water System: Wall Mounted
Produce ASTM type 1 water for critical analysis like HPLC, PCR, antibody production, cell culture media preparation or electrophoresis with the Sartorius arium™ Pro ASTM TYPE 1 - Ultrapure Water System.
Brand: Sartorius H2OPRO-UV-B
Description
The arium pro series consist of flexible systems with device configurations that can be specially custom-tailored to applications and feature an outstanding cost-benefit ratio. All systems meet and exceed ASTM Type 1 water quality and ensure the best, reproducible results. Per hour, an arium pro can deliver up to 120 l of ultrapure water with a conductivity of 0.055μS/cm (18.2 megaohm/cm at 25°C) and consistently high quality. When a Sartopore° 2 150 final filter is used, arium pro will deliver ultrapure water that is practically free of microorganisms.
- Mounts on wall
- Multiple configurations available based on application needs
- Optimized components and cartridge materials with topdown-flow technology
- Optional, integrated UV lamp (185/254nm) for eliminating bacteria and organics; controlled by reliable TOC monitoring
- Display with touch-activated functions and intuitive menu navigation
- Integrated real time monitoring and data display: feed conductivity, total water consumption, pressure regulator
- PIN-protected access to basic settings /service
- Convenient SD card slot
- System can be chosen based on daily work
- Guarantees a long service life and optimal water purification
- Reliable water quality ensures reproducible results
- Easy operation; no user manual needed
- Optimal control of constant water flow and consistent water quality without extra costs
- Prevents user operating errors
- Supports reliable recording of important data
Specifications
ASTM | |
<1 CFU/L | |
≤2 | |
18.2 Mohm.cm | |
Arium pro System with UV Lamp | |
2°C to 30°C | |
80% | |
Wall | |
4 to 10 | |
Adsorption by spherical activated Carbon, deionization, optional final particle and sterile filtration | |
35.6°F to 95°F | |
<1 | |
35 cm | |
49.2 cm | |
45.1 cm | |
27 to 29 kg | |
100/240 V |
0.055 | |
120 | |
Type 1 | |
<2 product water, <50 feed water | |
35.6°F to 86°F | |
Chromatographic | |
Wall mounting kit, dispenser unit with 2.8m extended cable, tubing and wall bracket | |
<1/mL | |
130 | |
5°C to 45°C | |
2°C to 35°C | |
13.7 in. | |
19.37 in. | |
17.75 in. | |
59.5 to 63.9 lb. | |
100/240 V 50/60 Hz | |
50/60 Hz |