Learn More
Sartorius arium™ RO-Module Cleaning Set
Effective removal of scaling and metal fouling
Brand: Sartorius H2O-CCS
60.29 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.32400kg
Includes:
- Alkaline cleaner HEDTA, ethanolamine, triethanolamine
- Acidic cleaner HEDTA, phosphoric acid, citric acid
Description
- Effective removal of scaling and metal fouling
- Elimination of organic compounds
- Dispersion of colloids
- Stable pH values
- Gentle on materials
Specifications
Water System | |
Arium™ RO-Module Cleaning Set | |
Alkaline cleaner HEDTA, Ethanolamine, Triethanolamine, Acidic cleaner HEDTA, Phosphoric Acid, Citric Acid |
Safety and Handling
- arium RO cleaning set
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : diammonium citrate