missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Biosart™ 100 Nutrient Media

Microbiological Nutrient Media for Filtration Units

£129.00 - £131.00

Specifications

Certifications/Compliance DIN, ISO
Content And Storage 4°C
Form Liquid
Manufacturing Origin United States
For Use With (Application) Microbiological testing of pharmaceuticals, food, beverages, Water and other liquids
View More Specs

Products 17
Product Code Brand Packaging Type Quantity Price Quantity & Availability  
Product Code Brand Packaging Type Quantity Price Quantity & Availability  
10516281
View Documents Promotion Details
Sartorius
16400-02----TT-K
Ampule 2.5 mL
£131.00
Pack of 50
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
11756025
View Documents Promotion Details
Sartorius
16400-02----CA-K
Ampule 2.5 mL
£131.00
Pack of 50
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
10747081
View Documents Promotion Details
Sartorius
16400-02----CE-K
Ampule 2.5 mL
£131.00
Pack of 50
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
10042301
View Documents Promotion Details
Sartorius
16400-02----KF-K
Ampule 2.5 mL
£131.00
Pack of 50
View alternative products
Save Up to 
»
Estimated Shipment: 14-10-2024
to see stock.
Add to basket
 
11766025
View Documents Promotion Details
Sartorius
16400-02----GS-K
Ampule 2.5 mL
£131.00
Pack of 50
View alternative products
Save Up to 
»
Estimated Shipment: 14-10-2024
to see stock.
Add to basket
 
11753693
View Documents Promotion Details
Sartorius
16400-02----EN-K
Ampule 2.5 mL
£131.00
Pack of 50
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
10072301
View Documents Promotion Details
Sartorius
16400-02----MF-K
Ampule 2.5 mL
£131.00
Pack of 50
View alternative products
Save Up to 
»
Estimated Shipment: 14-10-2024
to see stock.
Add to basket
 
10652602
View Documents Promotion Details
Sartorius
16400-02----MG-K
Ampule 2.5 mL
£129.00
Pack of 50
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
10544201
View Documents Promotion Details
Sartorius
16400-02----OS-K
Ampule 2.5 mL
£131.00
Pack of 50
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
10307601
View Documents Promotion Details
Sartorius
16400-02----RA-K
Ampule 2.5 mL
£131.00
Pack of 50
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
11786025
View Documents Promotion Details
Sartorius
16400-02----SB-K
Ampule 2.5 mL
£131.00
Pack of 50
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
10265421
View Documents Promotion Details
Sartorius
16400-02----WL-K
Ampule 2.5 mL
£131.00
Pack of 50
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
11762335
View Documents Promotion Details
Sartorius
16400-02----TZ-K
Ampule 2.5 mL
£131.00
Pack of 50
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
15932530
View Documents Promotion Details
Sartorius
16400-02----WZ-K
Ampule 2.5 mL
£131.00
Pack of 50
View alternative products
Save Up to 
»
Estimated Shipment: 14-10-2024
to see stock.
Add to basket
 
10178572
missing translation for 'viewDocuments' Promotion Details
Sartorius
16400-02----WN-K
Ampule 2.5 mL
£131.00
Pack of 50
View alternative products
missing translation for 'toSaveUpto' 
»
In Stock
to see stock.
Add to basket
 
10009920
missing translation for 'viewDocuments' Promotion Details
Sartorius
16400-02----TC-K
Ampule 2.5 mL
£131.00
Pack of 50
View alternative products
missing translation for 'toSaveUpto' 
»
In Stock
to see stock.
Add to basket
 
11776025
missing translation for 'viewDocuments' Promotion Details
Sartorius
16400-02----LS-K
Ampule 2.5 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
Description

Description

Biosart™ 100 Monitors have been specifically designed for microbiological testing of pharmaceuticals, food, beverages, water and other liquids. These sterile monitors with an incorporated membrane filter and cellulose pad are ready to use. Biosart™ 100 Nutrient Media for wetting the pad are available in individually sterilized, convenient plastic ampoules.Each box of Biosart™ 100 Nutrient Media contains 50 ampoules with sterile media, each with 2.5 ml and a lot certificate. If stored under proper conditions (+4°C), the culture media have a shelf life of 12 month (except for Endo, KF Strep, Lauryl Sulfate and Tergitol which have a 9-month shelf life). Biosart™ 100 Nutrient Media comply with international regulations and recommendations: International pharmacopoeias, DIN and ISO standards, the American Standards for Water and Foods, mineral water regulations, guidelines of the food and beverage industries.Within the scope of the quality assurance procedure and the stringent quality control standards every batch has passed Sartorius in-house tests of growth promotion, sterility, physical and technical parameters have been passed successfully. Biosart™100 Nutrient Media are convenient in use and eliminating the handling of glass ampoules.

  • presterilized media
  • certificate of quality for every batch
  • in compliance with international standards
  • consistent recovery
  • Ready-to-use
  • long shelf life

Specifications

Specifications

DIN, ISO
Liquid
Microbiological testing of pharmaceuticals, food, beverages, Water and other liquids
2.5 mL
4°C
United States
Ampule
Videos
SDS
Documents

Documents

Certificates