Learn More
Sartorius Cellulose Acetate Membrane Filters
Combines high flow rates and thermal stability with very low adsorption characteristics. Sartorius Cellulose Acetate Membrane Filters are excellently suited for use in pressure filtration devices. Non-sterile filters in a variety of diameters and pore sizes are bulk-packed to save money and space.
£78.20 - £829.00
Specifications
Autoclavable | Autoclavable |
---|---|
Color | White |
Material | Cellulose Acetate |
Sterility | Non-sterile |
For Use With (Application) | For use in pressure filtration devices; Sterile filtration of solution. |
Product Code | Brand | Pore Size | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Diameter (Metric) | Price | Quantity & Availability | ||||
10189130
|
Sartorius
11104--47------N |
0.8 μm | 47 mm |
£160.99
Pack of 100 |
In Stock
Log in to see stock. |
||||
10466551
|
Sartorius
11104--13------N |
0.8 μm | 13 mm |
£84.10
Pack of 100 |
In Stock
Log in to see stock. |
||||
10314021
|
Sartorius
11104--50------N |
0.8 μm | 50 mm |
£152.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
10271601
|
Sartorius
11104--25------N |
0.8 μm | 25 mm |
£117.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
10692392
|
Sartorius
11105--47------N |
0.65 μm | 47 mm |
£131.00
Pack of 100 |
Estimated Shipment: 19-08-2024
Log in to see stock. |
||||
10674532
|
Sartorius
11105--50------N |
0.65 μm | 50 mm |
£141.00
Pack of 100 |
Estimated Shipment: 02-09-2024
Log in to see stock. |
||||
10313401
|
Sartorius
11106-100------G |
0.45 μm | 100 mm |
£153.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
10154221
|
Sartorius
11106--30------N |
0.45 μm | 30 mm |
£114.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
10788701
|
Sartorius
11106--50------N |
0.45 μm | 50 mm |
£141.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
10796881
|
Sartorius
11106--13------N |
0.45 μm | 13 mm |
£78.20
Pack of 100 |
In Stock
Log in to see stock. |
||||
10475021
|
Sartorius
11106--25------N |
0.45 μm | 25 mm |
£98.10
Pack of 100 |
In Stock
Log in to see stock. |
||||
10739091
|
Sartorius
11106--90------G |
0.45 μm | 90 mm |
£144.00
Pack of 25 |
Estimated Shipment: 05-09-2024
Log in to see stock. |
||||
10788511
|
Sartorius
11106--47------N |
0.45 μm | 47 mm |
£218.78
Pack of 100 |
In Stock
Log in to see stock. |
||||
10422951
|
Sartorius
11107--13------N |
0.2 μm | 13 mm |
£128.00
Pack of 100 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||||
10709671
|
Sartorius
11107--25------N |
0.2 μm | 25 mm |
£98.10
Pack of 100 |
In Stock
Log in to see stock. |
||||
10622042
|
Sartorius
11107--30------N |
0.2 μm | 30 mm |
£114.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
10746981
|
Sartorius
11107--47------N |
0.2 μm | 47 mm |
£221.88
Pack of 100 |
In Stock
Log in to see stock. |
||||
10612561
|
Sartorius
11107--50------N |
0.2 μm | 50 mm |
£141.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
10528001
|
Sartorius
11107--90------G |
0.2 μm | 90 mm |
£157.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
10255511
|
Sartorius
12303-100------G |
1.2 μm | 100 mm |
£217.00
Pack of 25 |
Estimated Shipment: 03-09-2024
Log in to see stock. |
||||
10715902
|
Sartorius
12303--25------N |
1.2 μm | 25 mm |
£154.00
Pack of 100 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
10281981
|
Sartorius
12303--47------N |
1.2 μm | 47 mm |
£242.52
Pack of 100 |
In Stock
Log in to see stock. |
||||
10620183
|
Sartorius
12342--47------N |
5.0 μm | 47 mm |
£261.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
10173701
|
Sartorius
12342--25------N |
5.0 μm | 25 mm |
£164.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
10707751
|
Sartorius
11105--25------N |
0.65 μm | 25 mm |
£98.10
Pack of 100 |
Estimated Shipment: 26-09-2024
Log in to see stock. |
||||
12942661
|
Sartorius
11106--85------N |
0.45 μm | 85 mm |
£829.00
Pack of 100 |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
15912450
|
Sartorius
11104--90------N |
0.8 μm | 90 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12932661
|
Sartorius
11106--47------N |
0.45 μm | 47 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12922661
|
Sartorius
11106--25------N |
0.45 μm | 25 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10574191
|
Sartorius
11106--85------N |
0.45 μm | 85 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12902661
|
Sartorius
11107--47------N |
0.2 μm | 47 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Combines high flow rates and thermal stability with very low adsorption characteristics. Sartorius Cellulose Acetate Membrane Filters are excellently suited for use in pressure filtration devices. Non-sterile filters in a variety of diameters and pore sizes are bulk-packed to save money and space.
- Ideal for colony counting, particle testing, microscopy and sterility testing
- High-quality CA membrane filters for use in pressure filtration devices
- Available in pore sizes ranging from 0.2μm up to 5.0μm
- 0.2μm filters are the standard for sterile filtration of aqueous solutions, such as nutrient media, buffers and sera
- 5μm filters can be used as prefilters to retain coarse particles, while allowing microorganisms through
- White, non-sterile filters without grids in a variety of diameters (13 to 100mm)
- Can be sterilized by autoclaving at 121°C or 134°C; gamma irradiated to 25 kGy, dry heat or ethvlene oxide
The results of publications on adsorption are difficult to correlate, as mostly different test substances, conditions and detection methods were used, and the membranes were tested without previously being sterilized.
Specifications
Autoclavable | |
Cellulose Acetate | |
For use in pressure filtration devices; Sterile filtration of solution. | |
120 μm |
White | |
Non-sterile | |
Hydrophilic |