missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Vivaspin™ Filtrate Centrifugal Concentrators
Remove proteins from difficult to filter samples, even whole blood. Sartorius Vivaspin™ Filtrate performs centrifugal ultrafiltration against the direction of centrifugal force for fast and simple preparation of protein-free ultrafiltrates. PES and CTA membranes for 5000 to 300,000 Dalton MWCO.
£87.75 - £117.00
Specifications
Disposable | Reusable |
---|---|
Filter Type | Centrifugal Ultrafiltration |
Sample Volume | 0.5 to 2.5 mL |
Sterility | Non-sterile |
For Use With (Application) | Small volume centrifugal ultrafiltration of biological samples |
Product Code | Brand | MWCO | Material (Membrane) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | MWCO | Material (Membrane) | Price | Quantity & Availability | ||||
10323731
|
Sartorius
13239----------E |
10 kDa | Cellulose Triacetate |
£110.00
Pack of 12 |
In Stock
Log in to see stock. |
||||
10139470
|
Sartorius
13229----------E |
5 kDa | Cellulose Triacetate |
£117.00
Pack of 12 |
In Stock
Log in to see stock. |
||||
10212131
|
Sartorius
13249----------E |
20 kDa | Cellulose Triacetate |
£117.00
Pack of 12 |
In Stock
Log in to see stock. |
||||
10778821
|
Sartorius
13279----------E |
300 kDa | PES |
£117.00
Pack of 12 |
In Stock
Log in to see stock. |
||||
10333731
|
Sartorius
13269----------E |
100 kDa | PES |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11785525
|
Sartorius
13209----------E |
5000, 10,000, 20,000 and 100,000 Da | Cellulose Triacetate, PES |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Offered with multiple MWCOs and two membrane options:
- Cellulose triacetate (CTA) features high hydrophilicity and very low non-specific binding.
- Cast without a support that could bind filtered solutes, these membranes are ideal where high target molecule recovery from the filtrate is important.
- Polyethersulfone (PES) membranes exhibit low fouling characteristics, exceptional flux and broad pH compatibility.
- Have a uniform surface, devoid of hydrophobic or hydrophillic interactions, offering excellent protein recoveries for most solutions.
- Typical applications for Vivaspin™ Filtrate include:
- Sample preparation for free drug
- Metabolite or hormone analyses
- Sample deproteinization
- Separation of proteins from low molecular weight substances in biological samples
- Sample capacity of 0.5 to 2.5 mL
- May be used in fixed angle or swing bucket rotors, accepting standard conical or flat bottom 15 mL centrifuge tubes
- Typically filter 90% of the initial sample volume in 30 minutes
- Avoid premature membrane blockage by particle loaded samples
- Retrieve the ultrafiltrate immediately after centrifugation without any disassembly
Specifications
Reusable | |
0.5 to 2.5 mL | |
Small volume centrifugal ultrafiltration of biological samples | |
12 Pk. | |
93 mm |
Centrifugal Ultrafiltration | |
Non-sterile | |
2000xG (Fixed Angle Rotor); 2500xG (Swinging Bucket Rotor) | |
0.5 to 2.5 mL | |
14 mm |