Learn More
Sartorius™ Sartopore™ 2 150 Membrane Filter Capsule
Ensure excellent durability and flow rate capacities. The Sartorius Sartopore 2 150 ready-to-use membrane filter capsule provides a broad spectrum of chemical compatibility and an low amount of extractables.
Brand: Sartorius™ 5441307H4--CE
121.11 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.12600kg
Description
- Using a 4 in. quick connector, Sartopore 2 150 capsules can be connected to the outlet of an arium™ pro, 611 ultrapure water system or an arium™ dispense gun.
- Provides a broad spectrum of chemical compatibility from pH 1 to pH 14 with a low amount of extractables
- Capsules meet the requirements of current USP WFI quality standards inclusive of toxicity evaluations as per the USP plastic class VI test and test for extractables
- Unique pleated filter design combined with the highly asymmetric pore structure of the membrane allows for excellent flow rates and great durability
- Allows for easy ventilation of the capsule while simultaneously preventing product loss when placed at the farthes point upstream
- Eliminates the need to invest in cost-intensive filter holders for flat filters
- Hydrophilic, heterogeneous double-layered polyethersulfone membrane
- Final filter membrane: asymmetrical, polyethersulfone
- Prefilter membrane: asymmetrical, polyethersulfone
- Support fleece: polypropylene
- Inner core: polypropylene
- Connections: polypropylene
- Housing: polypropylene
- Bell assembly: polycarbonate
- Pore size combination: 0.45 μm + 0.2 μm
- Filtration area: 0.015 m2
- Inlet connection: 4 inch quick connector
- Outlet connection: 6 mm tube connector
- Max permitted differential pressure: 4 bar at 20°C, 2 bar at 80°C
- Max permitted counter pressure: 2 bar at 20°C
Specifications
HIMA and ASTM F-838-05, WFI | |
Particle removal and sterile filtration ofinjection solutions, therapeutics, biotechproducts, buffers, and ultrapure water. | |
Membrane Filter Capsule |