Learn More
Sartorius Disposable Plastic Syringes
Disposable Plastic Syringes that can be used with all syringe filter holders and 3-way valves.
Brand:  Sartorius 16644----------E
 View more versions of this product
Product Code. 10708591
  • £47.50 / Pack of 12
Edge
Explore more special offers
Description
Description
  • Male luer-lok connector
  • Comes with needle
  • Sterile
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Plastic
Syringe
5 mL
Sterile
For filter holders
Single-use
Cone