Learn More
Sartorius Entris™ Toploading Balances
When you need to get a heavy workload of repetitive applications done fast. Innocal Calibration Services avaliable on some models.
£800.00 - £1506.55
Specifications
Connection Type RS-232C, 25-pin
Stabilization Time 1.5 sec.
Type Toploading Balance
View More Specs
Products 16
Product Code Brand Capacity Readability Includes Price Quantity & Availability  
Product Code Brand Capacity Readability Includes Price Quantity & Availability  
15203923
View Documents
Sartorius
ENTRIS6202-1S
6.2 kg 10 mg External Calibration This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15213923
View Documents
Sartorius
ENTRIS4202-1S
4.2 kg 10 mg External Calibration This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15223923
View Documents
Sartorius
ENTRIS3202-1S
3.2 kg 10 mg External Calibration This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15233923
View Documents
Sartorius
ENTRIS2202-1S
2.2 kg 10 mg External Calibration This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15243923
View Documents
Sartorius
ENTRIS822-1S
820 g 10 mg External Calibration This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15253923
View Documents
Sartorius
ENTRIS8201-1S
8.2 kg 100 mg External Calibration This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15263923
View Documents
Sartorius
ENTRIS5201-1S
5.2 kg 100 mg External Calibration This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15273923
View Documents
Sartorius
ENTRIS2201-1S
2.2 kg 100 mg External Calibration This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15293903
View Documents
Sartorius
ENTRIS822I-1S
820 g 10 mg Internal Calibration This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15283903
View Documents
Sartorius
ENTRIS2202I-1S
2.2 kg 10 mg Internal Calibration This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15223913
View Documents
Sartorius
ENTRIS2201I-1S
2.2 kg 100 mg Internal Calibration This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15203913
View Documents
Sartorius
ENTRIS8201I-1S
8.2 kg 100 mg Internal Calibration This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15263903
View Documents
Sartorius
ENTRIS4202I-1S
4.2 kg 10 mg Internal Calibration This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15234946
View Documents
Sartorius
ENTRIS5201I-1S
5.2 kg 100 mg Internal Calibration This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15213913
View Documents
Sartorius
ENTRIS5201I-1S
5.2 kg 100 mg Internal Calibration This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15273903
View Documents
Sartorius
ENTRIS3202I-1S
3.2 kg 10 mg Internal Calibration This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • White-backlit, high-contrast display with 15mm digits; exceptionally easy to read under any room lighting conditions
  • New operating concept: easy configuration of the individual settings guided by short, menu-driven text prompts (choice of 5 languages) and simple navigation using cursor keys
  • RS-232C bidirectional interface port; optional Sartorius cable for connection of the balance to a USB port
  • Total ease of operation
  • All keys have positive click action
  • Clearly defined control panel
  • Monolithic weighing cell provides high thermal stability and extremely fast response times
  • Fast response time and display stability
  • Language-driven user interface
  • Unit toggle of up to 5 units
Applications
Percent weighing, unit toggle, animal weighing, net-total, calculation, density determination and counting
Innocal Calibration Services
Available in items containing "i-1SUS" in Manufacturer No.: 14-559-023, -024,- 025, -026, -027, -028, -029, -030.
Specifications
Specifications
RS-232C, 25-pin
1.5 sec.
Toploading Balance