missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Extra Thick Glass Fiber (GF) Pre-filters

Glass fiber prefilters, depth pre filters, for larger totally filterable volumes in clarification and sterile filtration.

Brand:  Sartorius 13430-124------K

Additional Details : Weight : 0.41200kg

 View more versions of this product

Product Code. 15932480

  • £140.00 / Pack of 50
Estimated Shipment: 23-08-2024
to see stock.
Add to basket

Description

Description

The major use of all three glass fibre filters is as a depth prefilter, placed directly on top of a membrane filter, whereby the prefilter diameter specified for the holder must be chosen. Larger diameters would intrude under the sealing ring of the holder and cause leakage. The standard type 13400 contains an acrlic binder. It has a high particle loading capacity, but for very dirty liquids the thicker type 13430 can be more effective. 13440 ia a finer , binder-free type and is recommended for the prefiltration of relative clean solutions such as tissue culture media.Serial filtration may be necessary for difficult-to-filter liquids such as serum.

Specifications

Specifications

Autoclavable
Disc
Non-sterile
Prefiltration of clean solutions such as tissue culture media
Hydrophilic
500°C
12.4 cm
White
Borosilicate Glass Microfiber
348 to 490mL/min.
13430
932°F
4.88 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers