Learn More
Sartorius Funnel
Reusable stainless steel funnels for microbial enumeration and particle counting.
Brand:  Sartorius 6981065
Product Code. 15694137
  • £369.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Specially designed for use in microbiological quality assurance, these three filter holders differ only in their capacity
  • The reusable funnels feature a special locking clamp that simplifies the positioning and removal of the membrane filter, ensuring secure lock-down
  • For traceability, each filter funnel has an individual serial number
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
Combisart System
Colony or particle counting
Stainless Steel
100 mL
Funnel
Closure clamp
4.7/5 cm