missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Funnels for Combisart™
Reusable stainless steel Combisart™ Funnels for microbial enumeration and particle counting.
£102.00 - £463.00
Specifications
Material | Stainless Steel |
---|---|
For Use With (Equipment) | Combisart Funnel |
Quantity | 1 Ea. |
Product Code | Brand | Product Type | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Includes | Price | Quantity & Availability | ||||
10243541
|
Sartorius
6981063 |
Funnel Lid | Small Central Air Port |
£102.00
Each |
In Stock
Log in to see stock. |
||||
10446741
|
Sartorius
6981002 |
Funnel | Locking Clamp |
£463.00
Each |
In Stock
Log in to see stock. |
||||
Description
- Specially designed for use in microbiological quality assurance, these three filter holders differ only in their capacity
- The reusable funnels feature a special locking clamp that simplifies the positioning and removal of the membrane filter, ensuring secure lock-down
- For traceability, each filter funnel has an individual serial number
Specifications
Stainless Steel | |
Combisart Funnel | |
1 Ea. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title