Learn More
Sartorius Gridded Nonsterile Cellulose Nitrate Membrane Filters: 0.45μm Pore Size
The Nonsterile Membrane Filters are made of cellulose nitrate (cellulose mixed ester), cellulose acetate or regenerated cellulose. They can be used for colony counting, particle testing and microscopy. Choose between gridded or non-gridded, membranes, either with or without hydrophobic edge.
£80.60 - £795.00
Specifications
Disposable | Single-use |
---|---|
Filter Type | Membrane Filter |
Material | Cellulose Nitrate |
Material (Membrane) | Cellulose Nitrate |
Pore Size | 0.45 μm |
Product Code | Brand | Diameter (Metric) | Grid Color | Color | Quantity | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Grid Color | Color | Quantity | Price | Quantity & Availability | ||
11730866
|
Sartorius
13006--47------R |
47 mm | Green | White | 1000 Pk. |
£795.00
Pack of 1000 |
Estimated Shipment: 24-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
10786781
|
Sartorius
13006--50------N |
50 mm | White | Gray | 100 Pk. |
£114.00
Pack of 100 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
10164081
|
Sartorius
13806--47------R |
47 mm | Dark Green | Green | 1000 Pk. |
£795.00
Pack of 1000 |
Estimated Shipment: 24-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
10403141
|
Sartorius
13806--50------N |
50 mm | Dark Green | Green | 100 Pk. |
£114.00
Pack of 100 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
10355031
|
Sartorius
13806--50------R |
50 mm | Dark Green | Green | 1000 Pk. |
£795.00
Pack of 1000 |
Estimated Shipment: 24-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
10007380
|
Sartorius
13906--50------R |
50 mm | Green | White | 1000 Pk. |
£795.00
Pack of 1000 |
Estimated Shipment: 24-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
10363441
|
Sartorius
11406--25------N |
25 mm | Black | White | 100 Pk. |
£80.60
Pack of 100 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
10588091
|
Sartorius
11406--47------N |
47 mm | Black | White | 100 Pk. |
£113.00
Pack of 100 |
In Stock
Log in to see stock. |
||
11730856
|
Sartorius
11406--47------R |
47 mm | Black | White | 1000 Pk. |
£790.00
Pack of 1000 |
Estimated Shipment: 24-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
10739281
|
Sartorius
13006--25------N |
25 mm | White | Gray | 100 Pk. |
£84.20
Pack of 100 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
10189320
|
Sartorius
13906--47------N |
47 mm | Green | White | 100 Pk. |
£127.00
Pack of 100 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
10633711
|
Sartorius
13906--50------N |
50 mm | Green | White | 100 Pk. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
If agar plates or absorbent pads to be wetted with liquid culture medium are used instead of Nutrient Pad Sets, Sartorius cellulose nitrate (cellulose ester) membrane filter are recommended. Naturally, the membrane filters must be free of microbes. For this purpose, they can be boiled or autoclaved. These membranes are offered in a choice of three different colors to suit the specific test application, and provide a high-contrast background to colonies or particles.These membrane filters are in accordance with the following norms: ISO 7704, ISO 7899-2, ISO 8199, ISO 9308-1 and EN 12780. In addition to this they have been manufactured for use especially at the same time with Sartorius Nutrient Pad Sets in accordance with many international regulations and recommendations: international pharmacopoeias, DIN and ISO Standards, the American Standards for Water and Foods, mineral water regulations, brewery guidelines, such as MEBAC or EBC, and recommendations of the food industry, such as LMBG, NCA and ICUMSA. All membranes are made of cellulose nitrate (cellulose ester), a material which assures effective retention with high flow rates and optimum colony growth. The printed grid divides the filtration area into 130 squares, each measuring 3.1 x 3.1 mm makes the counting easier, especially for higher bacteria counts and for micro-colonies, but does not influence the growth.
- Outstanding recovery rates for microorganisms
- 0.45 μm are acc. to ISO 7704
- Three different colors available
- Design: 25, 47 or 50 mm in diameter, white, grey or green and gridded
- Growth Promotion Test acc. to ISO 7704: no enhancement or inhibition by the grid lines, no enhancement or inhibition due to chemical extractables
Specifications
Single-use | |
Cellulose Nitrate | |
0.45 μm | |
For Colony Count, Sterility Testing, Particle Count, Microscopy |
Membrane Filter | |
Cellulose Nitrate | |
Non-sterile |