Learn More
Sartorius Gridded Sterile Cellulose Nitrate Membrane Filters: 0.45μm
Ensure quality control in microbiological filtering. Sartorius Gridded Sterile Cellulose Nitrate Membrane Filters, 0.45μm are single-packed membranes with colored grids for colony counting, particle testing and microscopy. The 0.45μm pore size is made in accordance with ISO 7704. Choose from two diameters and quantities in a range of filter/grid color combinations.
£127.00 - £1050.00
Specifications
Material | Cellulose Nitrate |
---|---|
Pore Size | 0.45 μm |
Sterility | Sterile |
For Use With (Application) | For colony count; sterility testing; particle count; microscopy |
Hydrophobicity | Hydrophilic |
Product Code | Brand | Diameter (Metric) | Type | Color | Grid Color | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Type | Color | Grid Color | Price | Quantity & Availability | ||
11700866
|
Sartorius
114H6--50----ACR |
50 mm | High Flow | White | Black |
£815.00
Pack of 1000 |
In Stock
Log in to see stock. |
||
11780636
|
Sartorius
139H6--50----ACN |
50 mm | High Flow | White | Green |
£130.00
Pack of 100 |
In Stock
Log in to see stock. |
||
11720876
|
Sartorius
139H6--47----ACR |
47 mm | High Flow | White | Green |
£880.00
Pack of 1000 |
In Stock
Log in to see stock. |
||
10738202
|
Sartorius
11406--50----ACR |
50 mm | Traditional | White | Black |
£885.00
Pack of 1000 |
In Stock
Log in to see stock. |
||
10243571
|
Sartorius
13006--50----ACR |
50 mm | Traditional | Gray | White |
£815.00
Pack of 1000 |
In Stock
Log in to see stock. |
||
10397921
|
Sartorius
13806--50----ACR |
50 mm | Traditional | Green | Dark Green |
£820.00
Pack of 1000 |
In Stock
Log in to see stock. |
||
10708131
|
Sartorius
13906--50----ACR |
50 mm | Traditional | White | Green |
£820.00
Pack of 1000 |
In Stock
Log in to see stock. |
||
10038080
|
Sartorius
13906--47----ACR |
47 mm | Traditional | White | Green |
£810.00
Pack of 1000 |
Estimated Shipment: 12-09-2024
Log in to see stock. |
||
10373291
|
Sartorius
11406--47----ACN |
47 mm | Traditional | White | Black |
£234.00
Pack of 100 |
In Stock
Log in to see stock. |
||
10236041
|
Sartorius
11406--47----ACR |
47 mm | Traditional | White | Black |
£1050.00
Pack of 1000 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
||
11780856
|
Sartorius
114H6--47----ACR |
47 mm | High Flow | White | Black |
£810.00
Pack of 1000 |
In Stock
Log in to see stock. |
||
11813220
|
Sartorius
114H6--47----ACN |
47 mm | High Flow | White | Black |
£127.00
Pack of 100 |
In Stock
Log in to see stock. |
||
10047950
|
Sartorius
13006--47----ACN |
47 mm | Traditional | Gray | White |
£165.00
Pack of 100 |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
10205521
|
Sartorius
13006--47----ACR |
47 mm | Traditional | Gray | White |
£880.00
Pack of 1000 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||
10282221
|
Sartorius
13006--50----ACN |
50 mm | Traditional | Gray | White |
£130.00
Pack of 100 |
In Stock
Log in to see stock. |
||
10109140
|
Sartorius
13806--47----ACN |
47 mm | Traditional | Green | Dark Green |
£152.00
Pack of 100 |
In Stock
Log in to see stock. |
||
10445401
|
Sartorius
13906--47----ACN |
47 mm | Traditional | White | Green |
£231.00
Pack of 100 |
In Stock
Log in to see stock. |
||
10633901
|
Sartorius
13906--50----ACN |
50 mm | Traditional | White | Green |
£130.00
Pack of 100 |
In Stock
Log in to see stock. |
||
10374103
|
Sartorius
139H6--47----ACN |
47 mm | High Flow | White | Black |
£152.00
Pack of 100 |
In Stock
Log in to see stock. |
||
Description
Ensure quality control in microbiological filtering. Sartorius Gridded Sterile Cellulose Nitrate Membrane Filters, 0.45μm are single-packed membranes with colored grids for colony counting, particle testing and microscopy. The 0.45μm pore size is made in accordance with ISO 7704. Choose from two diameters and quantities in a range of filter/grid color combinations.
Sterile, individually packaged filters for strict quality control- Pre-sterilized via gamma irradiation at 25kGray
- Ready-to-use to save preparatory time
- Avoids contamination of remaining filters in opened packs
- Filter identification and lot number are printed on each individual envelope
- Manufactured in accordance with ISO 7704, ISO 7899-2, ISO 8199, ISO 9308-1 and EN 12780
- Each membrane is checked to ensure:
- It is not damaged in any way
- It is positioned correctly with no slippage under the edge seal
- It is free of particles
- Each envelope has readable lettering
- Defined particle retention and outstanding, reliable recovery rates for microorganisms
- Cellulose nitrate filters (mixed cellulose ester) combine high flow rates with optimal colony growth
- The various-colored grids divide the filtration area into squares and provide the best possible color contrast
- Enables easy and reliable counting and identification of colonies and particles
- Does not influence the growth of bacteria and micro-colonies
- 0.45μm filters are manufactured in accordance with ISO 7704 standards
- Available in 47 and 50mm (1.9 or 2.0 in.) diameters
Specifications
Cellulose Nitrate | |
Sterile | |
Hydrophilic |
0.45 μm | |
For colony count; sterility testing; particle count; microscopy | |
Yes |