Learn More
Sartorius MD8 Airport Portable Air Sampler
MD8 Airport Portable Air Sampler for impaction measurements and background monitoring with gelatine membranes.
Brand:  Sartorius 16757
Product Code. 10638771
  • £6830.00 / Each
Explore more special offers
Description
Description
AirPort MD8 is the new air sampler for the pharmaceutical industry, the biotechnology, the food and beverage industry, for hospitals environmental care and for works safety. For guaranteeing reliable and exact measurement results AirPort MD8 uses the gelatine membrane filter method or the Impaction method with BACTair™.
  • Battery-powered and portable for universal use
  • Battery power level clearly indicated so constant performance during sampling is guaranteed
  • Ergonomic design and easy to clean
  • Flexible adjustment possibilities of the volume flow and the sample volume
  • Air Flow:
    • 10 L/min., 20 L/min., 30 L/min., 50 L/min.
    • 125 L/min. (only when BACTairTM culture media plates are used)
  • User friendly prompting with the option of five languages: English, French, German, Italian and Spanish
  • Parameters last used are stored even after automatic shut-off
  • The device can be calibrated on-site
  • CSA Compliant
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10 L/min., 20 L/min., 30 L/min., 50 L/min., 125 L/min.
Rechargeable NiMH
5 to 14 Watt depending of filter pressure drop
Portable Air Sampler
115/230 V
2.5 kg
Air Sampler, battery charger, operating instructions, calibration certificate.
CSA Compliant
For hospitals environmental care and for works safety
300 x 135 x 165 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Sartorius MD8 Airport Portable Air Sampler > MD8 Airport Portable Air Sampler; Airflow Rate: 10 L/min., 20 L/min., 30 L/min., 50 L/min., 125 L/min.