Learn More
Sartorius™ Microsart™ e.motion™ Membrane Dispenser
Versatile and flexible fully automated membrane filter dispenser for individually, sterile-packaged cellulose nitrate membrane filter discs
Brand:  Sartorius™ 16712
Product Code. 10520823
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Special feed roll technology makes each filter quickly and reliably accessible; avoids filter band slippage or even damaged membranes.
  • Compact and robust design
  • Works hands-free by an optical sensor (when sensor detects approaching tweezers) or touch button
  • Rapid and reliable transport due to sprocket feed roll technology
  • Controller prevents unwanted dispensing of several membrane filters at a time
  • Easy insertion of the filter band
  • Clear, compact design of the dispenser allows quick and easy cleaning
  • Low weight for easy transport
  • Interface port available so that other sensor systems can be connected to control the dispenser
Order Info
Accessory pedal switch available: 1ZE---0028. Membrane filters available separately.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CE and EMC, EN 50081-1 and -2, EN 50082-1 and -2, EN 61010
To prevent unwanted dispensing of several membrane filters
6.5 in.
2.9 kg
Membrane Filter
Membrane Filter Dispenser
16.5 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products