Learn More
Sartorius Polyethersulfone Membrane Filters
Polyethersulfone (PES) filters have a high internal porosity.
£140.00 - £306.50
Specifications
Material | PES |
---|---|
For Use With (Application) | Recommended for filtering biological and pharmaceutical solutions. |
Quantity | 100/Pk. |
Thickness (Metric) | 150 μm |
Product Code | Brand | Diameter (Metric) | Pore Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Pore Size | Price | Quantity & Availability | ||||
11753453
|
Sartorius
15406--25------N |
25 mm | 0.5 μm |
£140.00
Pack of 100 |
Estimated Shipment: 10-10-2024
Log in to see stock. |
||||
11763453
|
Sartorius
15406--47------N |
47 mm | 0.5 μm |
£209.00
Pack of 100 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||||
11733693
|
Sartorius
15407--25----MIN |
25 mm | 0.2 μm |
£140.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
11743693
|
Sartorius
15407--47----MIN |
47 mm | 0.2 μm |
£306.50
Pack of 100 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||||
11790686
|
Sartorius
15407--50----MIN |
50 mm | 0.2 μm |
£230.00
Pack of 100 |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
11713693
|
Sartorius
15458--25------N |
25 mm | 0.1 μm |
£143.00
Pack of 100 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
10275511
|
Sartorius
15458--47------N |
47 mm | 0.1 μm |
£209.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
10551094
|
Sartorius
15458--50------N |
50 mm | 0.1 μm |
£217.00
Pack of 100 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||||
Description
Polyethersulfone (PES) filters have a high internal porosity.
They perform well at high flux with an excellent throughput of aqueous solutions over the entire pH range of 1-14. Thanks to their low non-specific protein adsorption the PES type is recommended for filtering biological and pharmaceutical solutions.
Specifications
PES | |
Recommended for filtering biological and pharmaceutical solutions. | |
100/Pk. | |
150 μm |