Learn More
Sartorius Pump Heads for Vivaflow™ Masterflex Pump Models
Pump heads in various sizes for Vivaflow 50 and/or 200
£697.00 - £743.04
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
10161091
|
Sartorius
VFA013 |
Vivaflow™ 200 |
£743.04
Each |
In Stock
Log in to see stock. |
|||||
10121141
|
Sartorius
VFA012 |
Vivaflow™ 50 and 200 |
£697.00
Each |
Estimated Shipment: 19-09-2024
Log in to see stock. |
|||||
Description
- Masterflex, Easy-Load models
- Requires Masterflex Pump VFP002
Specifications
Pump Head |