Learn More
Sartorius Quantitative Grade 1289 Filter Papers
These qualitative filter papers are essentially used for analytical purposes and routine analyses, whenever no gravimetric analyses are required. They are wet-strengthened and can be used for pressure and vacuum filtration.
£10.20 - £59.00
Product Code | Brand | Diameter (Metric) | Grade | Particle Retention | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Grade | Particle Retention | Price | Quantity & Availability | |||
10471232
|
Sartorius
FT-3-208-070 |
70 mm | 1290 | 3 to 5 μm |
£10.80
Pack of 100 |
Estimated Shipment: 12-11-2024
Log in to see stock. |
|||
10387781
|
Sartorius
FT-3-208-055 |
55 mm | 1290 | 3 to 5 μm |
£10.20
Pack of 100 |
In Stock
Log in to see stock. |
|||
10778772
|
Sartorius
FT-3-207-240 |
240 mm | 1289 | 8 to 12 μm |
£59.00
Pack of 100 |
In Stock
Log in to see stock. |
|||
10480272
|
Sartorius
FT-3-207-125 |
125 mm | 1289 | 8 to 12 μm |
£18.60
Pack of 100 |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||
10032301
|
Sartorius
FT-3-207-185 |
185 mm | 1289 | 8 to 12 μm |
£36.30
Pack of 100 |
In Stock
Log in to see stock. |
|||
10205421
|
Sartorius
FT-3-207-110 |
110 mm | 1289 | 8 to 12 μm |
£14.50
Pack of 100 |
Estimated Shipment: 12-11-2024
Log in to see stock. |
|||
10133991
|
Sartorius
FT-3-207-070 |
70 mm | 1289 | 8 to 12 μm |
£10.80
Pack of 100 |
In Stock
Log in to see stock. |
|||
10767812
|
Sartorius
FT-3-207-090 |
90 mm | 1289 | 8 to 12 μm |
£11.10
Pack of 100 |
In Stock
Log in to see stock. |
|||
10490272
|
Sartorius
FT-3-207-150 |
150 mm | 1289 | 8 to 12 μm |
£24.20
Pack of 100 |
In Stock
Log in to see stock. |
|||
10226041
|
Sartorius
FT-3-207-055 |
55 mm | 1289 | 8 to 12 μm |
£10.20
Pack of 100 |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||
11761165
|
Sartorius
FT-3-207-050 |
50 mm | 1289 | 8 to 12 μm |
£13.25
Pack of 100 |
Estimated Shipment: 14-11-2024
Log in to see stock. |
|||
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.Specifications
Disc | |
100/Pk. | |
0.21 mm |