Learn More
Sartorius Sartolab™ 150v Vacuum Filtration Units
Optimised for the application in cell culture
Brand:  Sartorius 18080----------M
Product Code. 11728204
Description
Description
Sartolab 150v is a disposable, 0.2μm sterile, ready-to-use membrane filter capsule for highest convenience. Sartolab 150v capsules are made of a unique hydrophilic Polyethersulfone (PES) membrane providing outstanding total throughput, flow rate performance, low extractables and broadest chemical compatibility. Each 0.2 ++m bottle-top capsule filters 15 liters of aqueous solutions. The integrated on-off valve switch allows easy movement between receiving flasks.
  • Quality and reliability
  • Low protein binding
  • Fast flow and high throughput
  • Easy to use
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Flask Rack
Not Autoclavable
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Sartorius Sartolab™ 150v Vacuum Filtration Units > For 150V filtration unit; Pack of 3