missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ Sterility Testing Reusable System: Glass Cylinders
Shop All Sartorius Products
Click to view available options
:
Glass Cylinder, For 16523, 130mL
Description
Glass Cylinder - 14-555-944
- Spare part for reusable sterility test system
- For model 16522; 85mL
- Spare part for reusable sterility test system
- For 16523, 130mL capacity filter holder
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction