missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Reusable Sterility Testing System: Inlet Needles

Inlet Needles for use with Reusable Sterility Testing System.

Brand:  Sartorius 16964

 View more versions of this product

Product Code. 15982550

  • £77.70 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Inlet Needle - 14-555-844

  • Stainless Steel
  • For ampoule testing, infusion bottles
  • Pierces rubber septa on side to prevent blockage by rubber particles from piercing
  • Dia. outlet hose nipple, 5 mm
  • Size, long
Inlet Needle - 14-555-845
  • Stainless Steel
  • For ampoule testing, infusion bottles
  • Pierces rubber septa on side to prevent blockage by rubber particles from piercing
  • Dia. outlet hose nipple, 5 mm
  • Size, short

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Reusable
Reusable Sterility Testing System
Long Inlet Needles
5 mm
Inlet needle
Stainless Steel
0.2 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers