Learn More
Sartorius Sterisart™ NF Sterility Testing System
BIOPHARMA
Provides completely closed system sterility testing of pharmaceuticals, open containers, closed containers, pre-filled syringes, powders, and medical devices. Sartorius Sterisart™ NF Sterility Testing System is optimal for use with isolators.
Brand:  Sartorius 16478--------GBD
475.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11730975
  • £722.00 / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
International pharmacopoeias require the complete sterility of pharmaceutical products that are injected into the blood stream or that otherwise enter the body below the skin surface. As a manufacturer of such products, you are required to supply proof of sterility of the final product batch. Sterisart™ NF is a completely closed system for the sterility testing of pharmaceutical products. It is based on the membrane filter method, however it eliminates the procedure of manipulating the filters. By this the main risk of a secondary contamination and false positive results is eliminated. A peristaltic pump transfers the sample into the filtration units, and after rinsing, the filtration units are filled with media and used for incubation of the filters without any contact to the environment.
  • reliable Sartochem ™membrane
  • high retention of microbes
  • low adsorption
  • high mechanical stability
  • pre-installed color-coded tube clamps
  • easy-to-read graduated marks
  • user-friendly (several practical adapters available)
  • product-/lot number identification
  • gas-impermeable packaging for protection against sterilants
TRUSTED_SUSTAINABILITY
Specifications
Specifications
4 oz.
Sterility testing of pharmaceutical products, optimal for use in isolators
122°F
15.7 cm
0.45 μm
Medical devices
No Septum
120 mL
Containers bags with Luer Lock male connectors
50°C
Sterisart NF gamma with female Luer Lock connector
3 bar at 20°C
Liquids
Gamma irradiation
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Sartorius Sterisart™ NF Sterility Testing System > Sterisart NF gamma; Female Luer Lock connection