missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaflow™ 200 Module
BIOPHARMA

Vivaflow™ 200 Module is a plug and play laboratory cross flow cassette. Re-usable. Optimal for cell culture supernatant and virus concentration. High flow rate and recovery.

Brand:  Sartorius VF20H9

480.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15993030

  • £662.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Vivaflow™ 200 is a ready to use and reusable laboratory crossflow cassette for concentrating up to 5 L samples.
  • The membrane surface per device is 200 cm2.
  • One unit comes with all necessary accessories for running the device with a laboratory pump and a size 16 pump head.
  • For speeding up the concentration, two cassettes can be run simultaneously.
  • Vivaflow™ 200 is optimally suited for applications like cell culture supernatant concentration, virus concentration and water concentration e.g. in fish breeding facilities.
  • This economical product offers a standard of ease of use, reliability and flexibility unrivalled by any laboratory filtration system in the market.
  • The thin-channel, flip-flow recirculation path of the modules provides high crossflow velocities with minimum pump speed requirements.
  • A single 200 cm2 module typically concentrates 1 L by a factor of 50 in less than 30 min. Alternatively, two Vivaflow™ 200 Modules can be used in parallel for concentration of 5 L in under 75 minutes.
  • Near total recovery of the concentrate is achieved with a single final rinse.
  • Vivaflow™ 200 can be easily cleaned and re-used several times.
  • All accessories for sample concentration, except a peristaltic pump with a pump head for size 16 tubing, are included in the package for using a single cassette.
  • A 500 mL sealed diafiltration reservoir can be added to the system for rapid and convenient buffer exchange.

Equipment required:

  • The standard Vivaflow™ 200 package comes with tubing, pressure indicator, flow restrictor and high pressure pump tubing.
  • All you need is a peristaltic pump with a pump head capable of handling 6.4 mm OD tubing (size 16).

Running 2 Vivaflow™ 200 cassettes in parallel:

  • For speeding up the concentration, two Vivaflow™ 200 cassettes can easily be connected using a Y connector which is preassembeled with the correct tubing sizes and fittings (size 16 tubing for connecting the two Vivaflow™ 200 units, size 15 tubing for the pump).
  • Please note that you need a size 15 pump head for running two Vivaflows™ in parallel.
  • High flow rate and recovery.
  • Clear housing for process monitoring.
  • Ready to use single cassettes.
  • Easy set up of 2 cassettes for use in parallel.
  • Re-usable product.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ISO 9001
TFF/Crossflow
Hydrosart
Concentration desalting and buffer exchange
1 Ea.
Vivaflow 200 Module
5.3 mL
13.8 cm
Reusable
Acrylic
200 cm2
4 bar (58 psi)
60°C (140°F)
0.5 to 5 L
5.43 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.