missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Vivaflow™ 50R Crossflow Casettes
Unique, Plug and Play laboratory crossflow devices for concentrating samples like viruses and antibodies
£356.00 - £432.00
Specifications
Material | Acrylic |
---|---|
For Use With (Application) | Concentrating viruses and antibodies |
Includes | Pressure indicator, flow restrictor and size 16 PVC peristaltic tubing and fittings |
Sample Type | Protein, Virus, Nanoparticles |
Volume (Metric) | 100 mL to 1 L |
Includes: Pressure indicator, flow restrictor and size 16 PVC peristaltic tubing and fittings
Product Code | Brand | MWCO | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | MWCO | Price | Quantity & Availability | |||||
15983030
|
Sartorius
VF05H4 |
100 kDa |
£356.00
Each |
In Stock
Log in to see stock. |
|||||
15198279
|
Sartorius
VF05H2 |
30 kDa |
£432.00
Each |
In Stock
Log in to see stock. |
|||||
15178279
|
Sartorius
VF05H0 |
10 kDa |
£432.00
Each |
In Stock
Log in to see stock. |
|||||
Description
- Reusable device with low-binding Hydrosart™ (modified regenerated cellulose) membrane
- For concentrating up to 1L samples
- Active membrane area per device is 50cm2
- One unit comes with all necessary accessories for running device with laboratory pump and size 16 pump head
- For speeding up concentration, two cassettes can be run simultaniously
- Acrylic main housing and flow channel
- Polypropylene membrane support
- Nylon fittings and PVC tubing
- Compatible with 1M NaOH for cleaning and depyrogenation
- High flow rate and recovery
- L x W x H: 100x 24 x 100mm
- Pump flow rate: 200 to 400mL/min
- Maximum pressure: 45psi
- Maximum temperature: 60°C
- Antibody/recombinant protein concentration/diafiltration in biopharma research
- Concentration of small production lots of proteins for diagnostic purposes
- Concentration of viruses from cell culture supernatants or of environmental samples
- Nanoparticle concentration
Specifications
Acrylic | |
Pressure indicator, flow restrictor and size 16 PVC peristaltic tubing and fittings | |
100 mL to 1 L |
Concentrating viruses and antibodies | |
Protein, Virus, Nanoparticles |